Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114953 Supplier Invitrogen™ Supplier No. PA5114953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCDC34.

CCDC34 (Coiled-Coil Domain Containing 34) is a Protein Coding gene. Involved in spermatogenesis. Has a probable role in anterograde intraflagellar transport which is essential for the formation of sperm flagella.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC34
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CCDC34
Gene Accession No. Q3UI66, Q96HJ3
Gene Alias 2810027O19Rik; 4930522P10Rik; AI596202; CCDC34; coiled-coil domain containing 34; coiled-coil domain-containing protein 34; L15; NY-REN-41; NY-REN-41 antigen; RAMA3; Renal carcinoma antigen NY-REN-41; RGD1560583
Gene Symbols CCDC34
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCDC34(Accession Q96HJ3), corresponding to amino acid residues K300-K350.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362187, 68201, 91057
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.