Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC36 Antibody (OTI2D11), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272504IR
Description
CCDC36 Monoclonal specifically detects CCDC36 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CCDC36 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCDC36 | |
Full length recombinant protein of human CCDC36 (NP_835467) produced in E.coli. | |
0.1 mL | |
339834.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D11 | |
Western Blot, Immunohistochemistry | |
Cancer/testis antigen 74, coiled-coil domain containing 36, coiled-coil domain-containing protein 36, CT74FLJ25320 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction