Learn More
CCDC36 Mouse anti-Human, Clone: OTI2F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808389

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Required for DNA double-strand breaks (DSBs) formation in unsynapsed regions during meiotic recombination. Probably acts by forming a complex with MEI4 and REC114, which activates DSBs formation in unsynapsed regions, an essential step to ensure completion of synapsis. Not required for HORMAD1 functions in pairing-independent synaptonemal complex formation, ATR recruitment to unsynapsed axes, meiotic silencing of unsynapsed chromatin (MSUC) or meiotic surveillance.Specifications
CCDC36 | |
Monoclonal | |
Unconjugated | |
CCDC36 | |
CT74 | |
Mouse | |
Affinity Chromatography | |
RUO | |
339834 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IYA8 | |
CCDC36 | |
Full length human recombit protein of human CCDC36 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.