Learn More
Invitrogen™ CCDC39 Polyclonal Antibody

Description
Peptide sequence: QELSITQSLC KARERETESE EHFKAIAQRE LGRVKDEIQR LENEMASILE Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.
Specifications
Specifications
| Antigen | CCDC39 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Ccdc39 |
| Gene Accession No. | Q9UFE4 |
| Gene Alias | 4921507O14Rik; AI447426; AI844750; b2b1304Clo; b2b1735Clo; b2b2025.1Clo; CCDC39; CILD14; coiled-coil domain containing 39; coiled-coil domain-containing protein 39; D3Ertd789e; FAP59; I79_014225; RGD1306277 |
| Gene Symbols | Ccdc39 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.