Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC40 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306274PCP
Description
CCDC40 Polyclonal antibody specifically detects CCDC40 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CCDC40 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
coiled-coil domain containing 40, FLJ20753, FLJ32021, KIAA1640 | |
Produced in rabbits immunized with E. coli-derived Human CCDC40 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55036 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction