Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC42 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306102AF750
Description
CCDC42 Polyclonal specifically detects CCDC42 in Human samples. It is validated for Western Blot.Specifications
CCDC42 | |
Polyclonal | |
Western Blot | |
CCDC42A, coiled-coil domain containing 42, coiled-coil domain-containing protein 42A, FLJ32734 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CCDC42. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
146849 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction