Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569868 Supplier Invitrogen™ Supplier No. PA569868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

CCDC42 is required for sperm development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC42
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC42
Gene Accession No. 0
Gene Alias A530001H01Rik; CCDC42; CCDC42A; coiled-coil domain containing 42; coiled-coil domain-containing protein 42; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42; RGD1565925
Gene Symbols CCDC42
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat CCDC42.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303234
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.