Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC42 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578640
Description
Positive Control: 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCDC42 is required for sperm development.
Specifications
CCDC42 | |
Polyclonal | |
Unconjugated | |
CCDC42 | |
A530001H01Rik; CCDC42; CCDC42A; coiled-coil domain containing 42; coiled-coil domain-containing protein 42; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42; RGD1565925 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96M95 | |
CCDC42 | |
Recombinant protein encompassing a sequence within the center region of human CCDC42. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction