Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112096 Supplier Invitrogen™ Supplier No. PA5112096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with: Human, Mouse (Species predicted to react based on 100% sequence homology).

CCDC42 is required for sperm development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC42
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCDC42
Gene Accession No. Q96M95
Gene Alias A530001H01Rik; CCDC42; CCDC42A; coiled-coil domain containing 42; coiled-coil domain-containing protein 42; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42A; coiled-coil domain-containing protein 42; RGD1565925
Gene Symbols CCDC42
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human CCDC42.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.