Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC42B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571035 Supplier Invitrogen™ Supplier No. PA571035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC42B Polyclonal Antibody for Western Blot

CFAP73 may play a role in ciliary/flagellar motility by regulating the assembly and the activity of axonemal inner dynein arm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC42B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFAP73
Gene Accession No. A6NFT4
Gene Alias CCDC42B; CFAP73; cilia and flagella associated protein 73; Cilia- and flagella-associated protein 73; coiled-coil domain containing 42B; coiled-coil domain-containing protein 42B; MIA2
Gene Symbols CFAP73
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human CCDC42B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387885
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.