Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC42B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561765 Supplier Invitrogen™ Supplier No. PA561765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRLALQEKLS TKLPEQAEDH VPPVLRLLEK RQELVDADQA LQAQKEVFRT KTAALKQRWE QLEQKERELK GSFI Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 45%.

CFAP73 may play a role in ciliary/flagellar motility by regulating the assembly and the activity of axonemal inner dynein arm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC42B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP73
Gene Accession No. A6NFT4
Gene Alias CCDC42B; CFAP73; cilia and flagella associated protein 73; Cilia- and flagella-associated protein 73; coiled-coil domain containing 42B; coiled-coil domain-containing protein 42B; MIA2
Gene Symbols CFAP73
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC42B. Recombinant protein control fragment (Product #RP-100771).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387885
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.