Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC43 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097844AF700
Description
CCDC43 Polyclonal antibody specifically detects CCDC43 in Human samples. It is validated for Western BlotSpecifications
CCDC43 | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 43, coiled-coil domain-containing protein 43, FLJ31795 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain-containing protein 43 using the numbering given in entry NP_653210.2 (GeneID 124808). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
124808 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction