Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC43 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10097844
Description
CCDC43 Polyclonal specifically detects CCDC43 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CCDC43 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
coiled-coil domain containing 43, coiled-coil domain-containing protein 43, FLJ31795 | |
Rabbit | |
Affinity Purified | |
RUO | |
124808 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation | |
NP_653210.2 | |
CCDC43 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain-containing protein 43 using the numbering given in entry NP_653210.2 (GeneID 124808). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction