Learn More
CCDC43 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301838A
Description
The recommended shelf life for this product is 1 year from date of receipt.
CCDC43 is a coiled-coil domain containing protein of unknown function.Specifications
| CCDC43 | |
| Polyclonal | |
| Unconjugated | |
| CCDC43 | |
| FLJ31795 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124808 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96MW1 | |
| CCDC43 | |
| Between 194 and 224 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.