Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC43 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988299
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8299 Supplier Bioss Supplier No. BS8132R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC43 is a coiled-coil domain containing protein of unknown function.

Specifications

Antigen CCDC43
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC43
Gene Accession No. Q96MW1
Gene Alias 2610001E01Rik; Ccdc43; coiled-coil domain containing 43; Coiled-coil domain-containing protein 43; D11Ertd707e; FLJ31795; RGD1311929
Gene Symbols CCDC43
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC43.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124808
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.