Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC44 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18816225UL
Description
CCDC44 Polyclonal specifically detects CCDC44 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CCDC44 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
CCDC44translational activator of cytochrome c oxidase 1, clone HQ0477 PRO0477p, coiled-coil domain containing 44, Coiled-coil domain-containing protein 44, translational activator of mitochondrially encoded cytochrome c oxidase I, Translational activator of mitochondrially-encoded cytochrome c oxidase I | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TACO1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ERSRIFSKLCLNIRLAVKEGGPNPEHNSNLANILEVCRSKHMPKSTIETALKMEKSKDTYLLYEGRGPGGSSLLIE | |
25 μL | |
metabolism | |
51204 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction