Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531909
Description
Recommended positive controls: IMR32, MCF-7. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.
Specifications
| CCDC44 | |
| Polyclonal | |
| Unconjugated | |
| TACO1 | |
| 2310066I18Rik; CCDC44; clone HQ0477 PRO0477p; coiled-coil domain containing 44; coiled-coil domain-containing protein 44; coiled-coil domain-containing protein 44 {ECO:0000312; EMBL:AAI66900.1}; PRO0477; RGD1306784; Taco1; taco1 {ECO:0000250; Translational activator of cytochrome c oxidase 1; translational activator of cytochrome c oxidase 1 {ECO:0000250; translational activator of cytochrome c oxidase I; translational activator of mitochondrially encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase 1; Translational activator of mitochondrially-encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase I {ECO:0000250; UniProtKB:Q9BSH4} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51204 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.59 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BSH4 | |
| TACO1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CCDC44. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction