Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554474 Supplier Invitrogen™ Supplier No. PA554474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKFICDASSL HQVRKKLDSL GLCSVSCALE FIPNSKVQLA EPDLEQAAHL IQALSNHEDV IHVYDNI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.

TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC44
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TACO1
Gene Accession No. B2RYT9, Q8K0Z7, Q9BSH4
Gene Alias 2310066I18Rik; CCDC44; clone HQ0477 PRO0477p; coiled-coil domain containing 44; coiled-coil domain-containing protein 44; coiled-coil domain-containing protein 44 {ECO:0000312; EMBL:AAI66900.1}; PRO0477; RGD1306784; Taco1; taco1 {ECO:0000250; Translational activator of cytochrome c oxidase 1; translational activator of cytochrome c oxidase 1 {ECO:0000250; translational activator of cytochrome c oxidase I; translational activator of mitochondrially encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase 1; Translational activator of mitochondrially-encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase I {ECO:0000250; UniProtKB:Q9BSH4}
Gene Symbols TACO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC44. Recombinant protein control fragment (Product #RP-93330).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360645, 51204, 70207
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.