Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554479
Description
Immunogen sequence: ERSRIFSKLC LNIRLAVKEG GPNPEHNSNL ANILEVCRSK HMPKSTIETA LKMEKSKDTY LLYEGRGPGG SSLLIE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.
Specifications
CCDC44 | |
Polyclonal | |
Unconjugated | |
TACO1 | |
2310066I18Rik; CCDC44; clone HQ0477 PRO0477p; coiled-coil domain containing 44; coiled-coil domain-containing protein 44; coiled-coil domain-containing protein 44 {ECO:0000312; EMBL:AAI66900.1}; PRO0477; RGD1306784; Taco1; taco1 {ECO:0000250; Translational activator of cytochrome c oxidase 1; translational activator of cytochrome c oxidase 1 {ECO:0000250; translational activator of cytochrome c oxidase I; translational activator of mitochondrially encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase 1; Translational activator of mitochondrially-encoded cytochrome c oxidase I; translational activator of mitochondrially-encoded cytochrome c oxidase I {ECO:0000250; UniProtKB:Q9BSH4} | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51204 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSH4 | |
TACO1 | |
Recombinant protein corresponding to Human CCDC44. Recombinant protein control fragment (Product #RP-93328). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction