Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC46 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8300 Supplier Bioss Supplier No. BS8133R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC46 is a protein with filament, myosin tail and ATPase domains. Orthologs of the gene exist in mouse, rat and chimp.

Specifications

Antigen CCDC46
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEP112
Gene Accession No. Q8N8E3
Gene Alias 1700001M19Rik; 1700029K01Rik; 8430407H02Rik; AV043680; AV207351; Ccdc46; centrosomal protein 112; centrosomal protein 112kDa; Centrosomal protein of 112 kDa; CEP112; coiled-coil domain containing 46; Coiled-coil domain-containing protein 46; MACOCO; RGD1564168
Gene Symbols CEP112
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC46.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201134
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.