Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC47 Monoclonal Antibody (1-B9-5)
GREENER_CHOICE

Catalog No. PIMA532872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532872 Supplier Invitrogen™ Supplier No. MA532872
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCDC47 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. The CCDC47 gene maps to chromosome 17 at 17q23.3. Little is known about this single-pass membrane protein except that the coiled-coil domain is within the cytosolic domain near the carboxy terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC47
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1-B9-5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Ccdc47
Gene Accession No. Q96A33, Q9D024
Gene Alias 2610204L23Rik; Adipocyte-specific protein 4; asp4; C88307; calumin; ccd47; CCDC47; coiled-coil domain containing 47; coiled-coil domain-containing protein 47; GK001; MSTP041; PSEC0077; RGD1308813
Gene Symbols Ccdc47
Host Species Mouse
Immunogen Recombinant protein within Human CCDC47 aa 1-483.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57003, 67163
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.