Learn More
Invitrogen™ CCDC47 Polyclonal Antibody

Description
A suggested positive control is mouse heart tissue lysate. PA5-20856 can be used with blocking peptide PEP-0970.
Specifications
Specifications
| Antigen | CCDC47 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ccdc47 |
| Gene Accession No. | Q96A33, Q9D024 |
| Gene Alias | 2610204L23Rik; Adipocyte-specific protein 4; asp4; C88307; calumin; ccd47; CCDC47; coiled-coil domain containing 47; coiled-coil domain-containing protein 47; GK001; MSTP041; PSEC0077; RGD1308813 |
| Gene Symbols | Ccdc47 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.