Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560126 Supplier Invitrogen™ Supplier No. PA560126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLLEEKLVDV LQLLQRLRDL NISKRALGKI LLSTLDAFRD PTHEGRPSPA AILDALHQAL AACQLLRRQP SAPAS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC48
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFCC1
Gene Accession No. Q9HA90
Gene Alias C3orf73; Ccdc48; Ccdc48l1; coiled-coil domain containing 48; coiled-coil domain containing 48 like; coiled-coil domain containing 48-like1; coiled-coil domain-containing protein 48; EFCC1; EF-hand and coiled-coil domain containing 1; EF-hand and coiled-coil domain-containing protein 1; EF-hand domain-containing protein ENSP00000381169; EF-hand domain-containing protein ENSP00000381169 homolog
Gene Symbols EFCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC48. Recombinant protein control fragment (Product #RP-100119).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.