Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562261
Description
Immunogen sequence: MTYFGHFGGA NHAHTLGELE ACIAMLVEQL RTQGCGGRTL GTSEEEAELQ QKVEENEHLR LELQMVETER VRLS Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 58%.
The function of this protein remains unknown.
Specifications
CCDC48 | |
Polyclonal | |
Unconjugated | |
EFCC1 | |
C3orf73; Ccdc48; Ccdc48l1; coiled-coil domain containing 48; coiled-coil domain containing 48 like; coiled-coil domain containing 48-like1; coiled-coil domain-containing protein 48; EFCC1; EF-hand and coiled-coil domain containing 1; EF-hand and coiled-coil domain-containing protein 1; EF-hand domain-containing protein ENSP00000381169; EF-hand domain-containing protein ENSP00000381169 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HA90 | |
EFCC1 | |
Recombinant protein corresponding to Human CCDC48. Recombinant protein control fragment (Product #RP-100120). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction