Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC50 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6920R

Description
This protein is a bifunctional mRNA-capping enzyme exhibiting RNA 5′-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5′-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5′-diphosphate terminus.Specifications
CCDC50 | |
Polyclonal | |
Unconjugated | |
Ccdc50 | |
C3orf6; C3orf6h; Ccdc50; coiled-coil domain containing 50; Coiled-coil domain-containing protein 50; DFNA44; Protein Ymer; putative C3orf6 protein homolog; YMER; Ymer protein | |
Rabbit | |
Protein A | |
RUO | |
152137 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8IVM0 | |
Ccdc50 | |
KLH conjugated synthetic peptide derived from human CCDC50. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction