Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC51 Monoclonal Antibody (A8-F4-D1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA532875
Description
CCDC51 Monoclonal Antibody for Western Blot, IHC (P)

Specifications
CCDC51 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q96ER9 | |
CCDC51 | |
Recombinant protein with Human CCDC51 aa 311-411. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
A8-F4-D1 | |
Unconjugated | |
CCDC51 | |
5730568A12Rik; AI551049; CCDC51; coiled-coil domain containing 51; coiled-coil domain-containing protein 51; RGD1311466 | |
Mouse | |
Protein A | |
RUO | |
79714 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction