Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC52 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7989R

Description
Regulator required for centriole duplication, for proper bipolar spindle formation and chromosome congression in mitosis.Specifications
CCDC52 | |
Polyclonal | |
Unconjugated | |
SPICE1 | |
CCDC52; coiled-coil domain containing 52; Coiled-coil domain-containing protein 52; D16Ertd480e; RGD1310729; SPICE; Spice1; spindle and centriole associated protein 1; spindle and centriole protein; Spindle and centriole-associated protein; Spindle and centriole-associated protein 1 | |
Rabbit | |
Protein A | |
RUO | |
152185 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8N0Z3 | |
SPICE1 | |
KLH conjugated synthetic peptide derived from hu CCDC52. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction