Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100429 Supplier Invitrogen™ Supplier No. PA5100429
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CCDC53.

Acts as a component of the WASH core complex that functions as a nucleation-promoting factor (NPF) at the surface of endosomes, where it recruits and activates the Arp2/3 complex to induce actin polymerization, playing a key role in the fission of tubules that serve as transport intermediates during endosome sorting. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC53
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WASHC3
Gene Accession No. Q9CR27, Q9Y3C0
Gene Alias 2900091E11Rik; 5730495F03Rik; AD-016; Ccdc53; CGI-116; coiled-coil domain containing 53; coiled-coil domain-containing protein 53; RGD1563761; WASH complex subunit 3; WASH complex subunit CCDC53; WASHC3; x0009
Gene Symbols WASHC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human WASHC3(Accession Q9Y3C0), corresponding to amino acid residues V143-S193.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299707, 51019, 67282
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.