Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC53 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558348 Supplier Invitrogen™ Supplier No. PA558348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMGSGIDLTK VPAIQQKRTV AFLNQFVVHT VQFLNRFSTV CEEKLADLSL RIQQIETTLN ILDAKLSSIP GLDDVTVEVS P Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 95%.

Acts as a component of the WASH core complex that functions as a nucleation-promoting factor (NPF) at the surface of endosomes, where it recruits and activates the Arp2/3 complex to induce actin polymerization, playing a key role in the fission of tubules that serve as transport intermediates during endosome sorting. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC53
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WASHC3
Gene Accession No. Q9Y3C0
Gene Alias 2900091E11Rik; 5730495F03Rik; AD-016; Ccdc53; CGI-116; coiled-coil domain containing 53; coiled-coil domain-containing protein 53; RGD1563761; WASH complex subunit 3; WASH complex subunit CCDC53; WASHC3; x0009
Gene Symbols WASHC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC53. Recombinant protein control fragment (Product #RP-97726).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.