Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548208
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548208 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548208 Supplier Invitrogen™ Supplier No. PA548208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with monkey based on sequence homology.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC54
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CCDC54
Gene Accession No. Q8NEL0
Gene Alias 1700007N18Rik; AI644412; BB013989; Ccdc54; coiled-coil domain containing 54; coiled-coil domain-containing protein 54; NYD-SP17; RGD1559779; SP17; sperm protein 17; testes development-related NYD-SP17; testicular tissue protein Li 33; testis development protein NYD-SP17
Gene Symbols CCDC54
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 91-119 amino acids from the Central region of human CCDC54.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.