Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC54 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988302
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8302 Supplier Bioss Supplier No. BS8135R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CCDC54
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC54
Gene Accession No. Q8NEL0
Gene Alias CCDC54; coiled-coil domain containing 54; Coiled-coil domain-containing protein 54; NYD-SP17; SP17; sperm protein 17; testes development-related NYD-SP17; testicular tissue protein Li 33; Testis development protein NYD-SP17
Gene Symbols CCDC54
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC54.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84692
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.