Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554766 Supplier Invitrogen™ Supplier No. PA554766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEEEVKLKAQ VARSQQDIER YKQQLSLAVE RERSLERDQV QLGLDWQRRC DDIERDQIQK SEALIQGLSM AKSQVAAKL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

CCDC57 (Coiled-Coil Domain Containing 57) is a Protein Coding gene. Diseases associated with CCDC57 include Uterine Benign Neoplasm and Reproductive Organ Benign Neoplasm. An important paralog of this gene is CCHCR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC57
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC57
Gene Accession No. Q2TAC2
Gene Alias CCDC57; coiled-coil domain containing 57; coiled-coil domain-containing protein 57
Gene Symbols CCDC57
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC57. Recombinant protein control fragment (Product #RP-93440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.