Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC58 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561587 Supplier Thermo Scientific Supplier No. PA561587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSGGVNCEEF AEFQELLKVM RTIDDRIVHE LNTTVPTASF AGKIDASQTC KQLYESLMAA HASRDRVI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.

CCDC58 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC58
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC58
Gene Accession No. Q4VC31
Gene Alias A930007B11Rik; AI413631; CCDC58; coiled-coil domain containing 58; coiled-coil domain-containing protein 58; RGD1564582
Gene Symbols CCDC58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC58
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.