Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553905 Supplier Invitrogen™ Supplier No. PA553905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKKIQALQKE KETLAVNYEK EEEFLTNELS RKLMQLQHEK AELEQHLEQE QEFQVNKLMK KIKKLENDTI SKQLTLEQLR REKI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC6
Gene Accession No. Q16204
Gene Alias 2810012H18Rik; AA536681; AA960498; AW061011; CCDC6; coiled-coil domain containing 6; coiled-coil domain-containing protein 6; D10S170; H4; Papillary thyroid carcinoma-encoded protein; Protein H4; PTC; TPC; TST1
Gene Symbols CCDC6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC6. Recombinant protein control fragment (Product #RP-92555).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.