Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988188
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8188 Supplier Bioss Supplier No. BS7990R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a coiled-coil domain-containing protein. The encoded protein is ubiquitously expressed and may function as a tumor suppressor. A chromosomal rearrangement resulting in the expression of a fusion gene containing a portion of this gene and the intracellular kinase-encoding domain of the ret proto-oncogene is the cause of thyroid papillary carcinoma.

Specifications

Antigen CCDC6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC6
Gene Accession No. Q16204
Gene Alias 2810012H18Rik; AA536681; AA960498; AW061011; CCDC6; coiled-coil domain containing 6; Coiled-coil domain-containing protein 6; D10S170; H4; Papillary thyroid carcinoma-encoded protein; Protein H4; PTC; TPC; TST1
Gene Symbols CCDC6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8030
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.