Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119038
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119038 200 μL
PIPA5122148 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119038 Supplier Invitrogen™ Supplier No. PA5119038
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC60 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

CCDC60 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC60
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CCDC60
Gene Accession No. Q8C4J0
Gene Alias AU018954; C130098D09; Ccdc60; coiled-coil domain containing 60; coiled-coil domain-containing protein 60; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 60
Gene Symbols CCDC60
Host Species Rabbit
Immunogen Recombinant protein Coiled Coil Domain Containing Protein 60. The antigen corresponds to amino acid range 1-186 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269693
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.