Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559140 Supplier Invitrogen™ Supplier No. PA559140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTGNFKQFNI FCHMLESALT QSSESVTLDL LTYTDLESLR NRKMGGRPGS LAPRSAQLNS KRYLILIYSV EFDRIHYPLP LPYQGKPDPV VLQGIIRSLK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

CCDC61 is a protein coding gene. Gene ontology (GO) annotation include centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC61
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC61
Gene Accession No. Q9Y6R9
Gene Alias C530028I08Rik; Ccdc61; coiled-coil domain containing 61; coiled-coil domain-containing protein 61; Gm159; RGD1307390
Gene Symbols CCDC61
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC61. Recombinant protein control fragment (Product #RP-98206).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.