Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587827 Supplier Invitrogen™ Supplier No. PA587827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPSPTGGRA LRFDPTAFVK AKERKQREIQ MKQQQRNRLG SGGSGDGPSV SWSRQTQPPA ALTGRGDAPN RSRNRSSSVD SFRSRCSSAS SCSDLEDFSE SLSRGGHRRR GKPPSPTPWS GSNMKSPPVE RSHHQKSLAN SGGWVPIKEY SSEHQAADMA EIDARLKALQ EYMNRLDMRS; Positive Samples: Jurkat.

CCDC61 is a protein coding gene. Gene ontology (GO) annotation include centrosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC61
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCDC61
Gene Accession No. Q9Y6R9
Gene Alias C530028I08Rik; Ccdc61; coiled-coil domain containing 61; coiled-coil domain-containing protein 61; Gm159; RGD1307390
Gene Symbols CCDC61
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 352-531 of human CCDC61 (NP_0010738711).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729440
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.