Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC62 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525081

Catalog No. PIPA525081


Only null left
Add to Cart

Description

Description

CCDC62 Polyclonal Antibody for Western Blot, IHC (P)

Nuclear receptor coactivator that can enhance preferentially estrogen receptors ESR1 and ESR2 transactivation. Modulates also progesterone/PGR, glucocorticoid/NR3C1 and androgen/AR receptors transactivation, although at lower level; little effect on vitamin D receptor/VDR. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCDC62
Polyclonal
Unconjugated
CCDC62
aaa-protein; AI661708; cancer/testis antigen 109; Ccdc62; coiled-coil domain containing 62; coiled-coil domain-containing protein 60; coiled-coil domain-containing protein 62; CT109; ERAP75; Gm150; Protein aaa; Protein TSP-NY; testis-specific protein TSP-NY; TSP-NY
Rabbit
Antigen affinity chromatography, Protein A
RUO
84660
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6P9F0
CCDC62
KLH conjugated synthetic peptide between 651-679 amino acids from the C-terminal region of human CCDC62.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.