Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563609 Supplier Invitrogen™ Supplier No. PA563609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERSEISCCQ KNEACLGESG MCDSKCCHPS NFIIEAPGHM SDVEWMSIFK PSKMQRIVRL KSGCTCSESI CGTQHDSPAS ELIAIQDSHS LGSSKSALRE DETES Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 64%.

Nuclear receptor coactivator that can enhance preferentially estrogen receptors ESR1 and ESR2 transactivation. Modulates also progesterone/PGR, glucocorticoid/NR3C1 and androgen/AR receptors transactivation, although at lower level; little effect on vitamin D receptor/VDR. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC62
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC62
Gene Accession No. Q6P9F0
Gene Alias aaa-protein; AI661708; cancer/testis antigen 109; Ccdc62; coiled-coil domain containing 62; coiled-coil domain-containing protein 60; coiled-coil domain-containing protein 62; CT109; ERAP75; Gm150; Protein aaa; Protein TSP-NY; testis-specific protein TSP-NY; TSP-NY
Gene Symbols CCDC62
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC62. Recombinant protein control fragment (Product #RP-97747).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.