Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC62 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989767
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9767 Supplier Bioss Supplier No. BS13807R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein has not been specifically defined.

Specifications

Antigen CCDC62
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene CCDC62
Gene Accession No. Q6P9F0
Gene Alias aaa-protein; AI661708; cancer/testis antigen 109; Ccdc62; coiled-coil domain containing 62; coiled-coil domain-containing protein 60; Coiled-coil domain-containing protein 62; CT109; ERAP75; Gm150; Protein aaa; Protein TSP-NY; testis-specific protein TSP-NY; TSP-NY
Gene Symbols CCDC62
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC62.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84660
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.