Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566367 Supplier Invitrogen™ Supplier No. PA566367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRLQLWEAYCQ VRYLCSHLRG NDSADSAVST DSSMDESSET SSAKDVPAGS LRTALNELKR LIQSIVDGME PTVT Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

CCDC64 is a component of secretory vesicle machinery in developing neurons that acts as a regulator of neurite outgrowth. It regulates the secretory vesicle transport by controlling the accumulation of Rab6-containing secretory vesicles in the pericentrosomal region restricting anterograde secretory transport during the early phase of neuronal differentiation, thereby inhibiting neuritogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC64
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BICDL1
Gene Accession No. Q6ZP65
Gene Alias 2210403N09Rik; Bicaudal D related 1; bicaudal D-related protein 1; bicaudal-D-related protein 1; BICD family like cargo adaptor 1; BICD family-like cargo adapter 1; Bicdl1; Bicdr1; BICDR-1; BICD-related protein 1; Ccdc64; CCDC64A; coiled-coil domain containing 64; coiled-coil domain-containing protein 64A; H_267D11.1; RGD1310292; WUGSC:H_267D11.1
Gene Symbols BICDL1
Host Species Rabbit
Immunogen Recombinant Human CCDC64. Recombinant protein control fragment (Product #RP-105169).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.