Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC65 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP18196825UL
Description
CCDC65 Polyclonal specifically detects CCDC65 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CCDC65 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q8IXS2 | |
CCDC65 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SSVLTPKEQEGIQKNNLEELTEELTKVMVDYIGMENFWKRYNKVKLEQLSLQHRRAQLLDINGKLREMLKQYLDGISVSDEVLSQ | |
Affinity Purified | |
RUO | |
85478 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
coiled-coil domain containing 65, coiled-coil domain-containing protein 65, FLJ25663, FLJ35732, NYD-SP28, Testis development protein NYD-SP28 | |
Rabbit | |
57 kDa | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction