Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558424 Supplier Invitrogen™ Supplier No. PA558424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSVLTPKEQE GIQKNNLEEL TEELTKVMVD YIGMENFWKR YNKVKLEQLS LQHRRAQLLD INGKLREMLK QYLDGISVSD EVLSQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

May play a role in motile cilia function, possibly by acting on the assembly of the nexin-dynein regulatory complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC65
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC65
Gene Accession No. Q8IXS2
Gene Alias 4933417K04Rik; AI428139; AU020178; CCDC65; CFAP250; CILD27; coiled-coil domain containing 65; coiled-coil domain-containing protein 65; Drc2; Dynein regulatory complex subunit 2; FAP250; NYD-SP28; Testis development protein NYD-SP28
Gene Symbols CCDC65
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC65. Recombinant protein control fragment (Product #RP-97766).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85478
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.