Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC65 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988277
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8277 Supplier Bioss Supplier No. BS8100R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in motile cilia function, possibly by acting on the assembly of the nexin-dynein regulatory complex.

Specifications

Antigen CCDC65
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC65
Gene Accession No. Q8IXS2
Gene Alias 4933417K04Rik; AI428139; AU020178; CCDC65; CFAP250; CILD27; coiled-coil domain containing 65; Coiled-coil domain-containing protein 65; DRC2; Dynein regulatory complex subunit 2; FAP250; NYD-SP28; Testis development protein NYD-SP28
Gene Symbols CCDC65
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC65.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85478
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.