Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546125 Supplier Invitrogen™ Supplier No. PA546125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PLRTKTGHIL KSTQDTCIGS EKLLQKKPVG SETSQAKGEK NGMTFSSTKD Sequence homology: Cow: 79%; Dog: 79%; Horse: 79%; Human: 100%.

Microtubule-binding protein required for ciliogenesis (PubMed:28235840). May function in ciliogenesis by mediating the transport of proteins like BBS4 to the cilium, but also through the organization of the centriolar satellites (PubMed:28235840). Plays a role in retina morphogenesis and/or homeostasis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC66
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC66
Gene Accession No. A2RUB6
Gene Alias CCDC66; coiled-coil domain containing 66; coiled-coil domain-containing protein 66; DKFZp686C0433; hypothetical protein LOC285331
Gene Symbols CCDC66
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC66 (aa 45-94).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285331
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.