Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC66 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140074
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QKGHDTSRLI KNLGVDTIQM EYNASNISNS RHDSDEISGK MNTYMNSTTS KKDTGVQTDD LNIGIFTNAE SHCGSLMERD ITNCSSPEIS AELI.
Microtubule-binding protein required for ciliogenesis (PubMed:28235840). May function in ciliogenesis by mediating the transport of proteins like BBS4 to the cilium, but also through the organization of the centriolar satellites (PubMed:28235840). Plays a role in retina morphogenesis and/or homeostasis. [UniProt].
Specifications
| CCDC66 | |
| Polyclonal | |
| Unconjugated | |
| CCDC66 | |
| CCDC66; coiled-coil domain containing 66; coiled-coil domain-containing protein 66; DKFZp686C0433; hypothetical protein LOC285331 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285331 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A2RUB6 | |
| CCDC66 | |
| Recombinant protein corresponding to Human CCDC66. Recombinant protein control fragment (Product #RP-108888). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction