Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC67 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988280
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8280 Supplier Bioss Supplier No. BS8105R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC67 (deuterosome assembly protein 1) is a key structural component of the deuterosome, a structure that promotes de novo centriole amplification in multiciliated cells. Deuterosome-mediated centriole amplification occurs in terminally differentiated multiciliated cells and can generate more than 100 centrioles. Probably sufficient for the specification and formation of the deuterosome inner core. The protein interacts with CEP152 and recruits PLK4 to activate centriole biogenesis

Specifications

Antigen CCDC67
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Deup1
Gene Accession No. Q05D60
Gene Alias 4933401K09Rik; Ccdc67; coiled-coil domain containing 67; Coiled-coil domain-containing protein 67; Deup1; deuterosomal protein 1; Deuterosome assembly protein 1; deuterosome protein 1
Gene Symbols Deup1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC67.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 159989
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.