Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC68 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24751925UL
Description
CCDC68 Polyclonal specifically detects CCDC68 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CCDC68 | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
coiled-coil domain containing 68, coiled-coil domain-containing protein 68, CTCL tumor antigen se57-1, CTCL-associated antigen se57-1, cutaneous T-cell lymphoma associated antigen, Cutaneous T-cell lymphoma-associated antigen se57-1, FLJ25368, SE57-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
80323 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CCDC68 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QKLKQHVENLNQVAEKLEEKHSQITELENLVQRMEKEKRTLLERKLSLENKLLQLKSSATYGKSCQDLQREISILQEQISHLQF | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction