Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565748 Supplier Invitrogen™ Supplier No. PA565748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPHELGPLNGD TAITVQLCAS EEAERHQKDI TRILQQHEEE KKKWAQQVEK ERELELRDRL DEQQRVLEGK NEEALQVLRA Highest antigen sequence identity to the following orthologs - mouse 30%, rat 30%.

May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC69
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC69
Gene Accession No. A6NI79
Gene Alias 2210021E03Rik; CCDC69; coiled-coil domain containing 69; coiled-coil domain-containing protein 69; D11Ertd461e; RGD1562251
Gene Symbols CCDC69
Host Species Rabbit
Immunogen Recombinant Human CCDC69. Recombinant protein control fragment (Product #RP-104955).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.